ENAH11DD2216 Enamine

N-ethyl-2,6-difluorobenzamide

  • MDL number MFCD03551077
  • Cas Number : 401587-34-6
  • Molecular Weight : 185.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-ethyl-2,6-difluorobenzamide
Inchi Code 1S/C9H9F2NO/c1-2-12-9(13)8-6(10)4-3-5-7(8)11/h3-5H,2H2,1H3,(H,12,13)
Inchi Key UQFMJCCMJOKHMH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1724156

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1724156
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart