ENAH11DD221C Enamine

9-methoxy-3,4,5,6,7,8-hexahydro-2H-azonine

  • Cas Number : 1086265-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724169
Availability details for
Quantity
  • Availability
  • Add to Cart