ENAH11DD2483 Enamine

sodium 3-(2,5-dichlorobenzenesulfonyl)propanoate

  • MDL number MFCD31666774
  • Cas Number : 2228826-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724218
Availability details for
Quantity
  • Availability
  • Add to Cart