ENAH11DD2487 Enamine

2,5-dioxopyrrolidin-1-yl (2Z)-3-(1H-imidazol-1-yl)but-2-enoate

  • MDL number MFCD31666591
  • Cas Number : 2229681-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724769
Availability details for
Quantity
  • Availability
  • Add to Cart