ENAH11DD24BA Enamine

{3,3-dimethyl-2-oxabicyclo[2.1.1]hexan-1-yl}methanol

  • MDL number MFCD31666553
  • Cas Number : 2228305-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725764
Availability details for
Quantity
  • Availability
  • Add to Cart