ENAH11DD24BB Enamine

methyl 2-amino-5-methylhexanoate hydrochloride

  • MDL number MFCD31666671
  • Cas Number : 50833-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725769
Availability details for
Quantity
  • Availability
  • Add to Cart