ENAH11DD24BD Enamine

N-(3-methoxypropyl)-2-(piperazin-1-yl)acetamide dihydrochloride

  • MDL number MFCD31666621
  • Cas Number : 2229561-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725781
Availability details for
Quantity
  • Availability
  • Add to Cart