ENAH11DD24C9 Enamine

(Z)-(1-amino-2-{[(tert-butoxy)carbonyl]amino}ethylidene)amino 2-phenoxyacetate

  • MDL number MFCD31690886
  • Cas Number : 2229669-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726017
Availability details for
Quantity
  • Availability
  • Add to Cart