ENAH11DD26E0 Enamine

(4R)-6-fluoro-3,4-dihydrospiro[1-benzopyran-2,1'-cyclobutane]-4-amine hydrochloride

  • MDL number MFCD28902010
  • Cas Number : 2227876-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724208
Availability details for
Quantity
  • Availability
  • Add to Cart