ENAH11DD26F3 Enamine

2-(difluoromethyl)-3-methylpyridine

  • MDL number MFCD25476557
  • Cas Number : 1805301-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725742

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P270,P271,P280,P301+P312,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P370+P378,P403+P233,P403+P235,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-1725742
pictograms GHS02,GHS07
hstatements H226,H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart