ENAH11DD26F6 Enamine

1-({[(tert-butoxy)carbonyl]amino}methyl)-3,3-dimethyl-2-oxabicyclo[2.1.1]hexane-4-carboxylic acid

  • MDL number MFCD31666650
  • Cas Number : 2228327-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725750
Availability details for
Quantity
  • Availability
  • Add to Cart