ENAH11DD26FE Enamine

4-(propan-2-yl)-2-[(2S)-pyrrolidin-2-yl]-1,3-oxazole dihydrochloride

  • MDL number MFCD31690773
  • Cas Number : 2227829-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725777
Availability details for
Quantity
  • Availability
  • Add to Cart