ENAH11DD2700 Enamine

methyl 2-chloro-2-(3,5-difluorophenyl)acetate

  • MDL number MFCD31665346
  • Cas Number : 1697762-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724938
Availability details for
Quantity
  • Availability
  • Add to Cart