ENAH11DD270A Enamine

[(2S)-1-(benzenesulfonyl)pyrrolidin-2-yl]methanol

  • MDL number MFCD11611368
  • Cas Number : 122030-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725841

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes ,Xi
MSDS https://www.enaminestore.com/catalog/EN300-1725841
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart