ENAH11DD270F Enamine

1-methyl-1,3,8-triazaspiro[4.5]decan-4-one hydrochloride

  • MDL number MFCD22570255
  • Cas Number : 1007-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725878
Availability details for
Quantity
  • Availability
  • Add to Cart