ENAH11DD2712 Enamine

3-(pyridin-3-yl)aniline dihydrochloride

  • MDL number MFCD11052344
  • Cas Number : 1235380-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725884
Availability details for
Quantity
  • Availability
  • Add to Cart