ENAH11DD2763 Enamine

5-amino-2,2-difluorospiro[2.3]hexane-1-carboxylic acid hydrochloride

  • MDL number MFCD31666664
  • Cas Number : 2228648-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726162
Availability details for
Quantity
  • Availability
  • Add to Cart