ENAH11DD2764 Enamine

ethyl 2-oxo-decahydronaphthalene-4a-carboxylate

  • MDL number MFCD31666801
  • Cas Number : 93088-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726163
Availability details for
Quantity
  • Availability
  • Add to Cart