ENAH11DD2775 Enamine

(2E)-3-(2-methylpyridin-3-yl)prop-2-enoic acid hydrochloride

  • MDL number MFCD29041199

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726228
Availability details for
Quantity
  • Availability
  • Add to Cart