ENAH11DD2776 Enamine

(2E)-3-[4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)phenyl]prop-2-enoic acid

  • MDL number MFCD02682306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726229
Availability details for
Quantity
  • Availability
  • Add to Cart