ENAH11DD277F Enamine

2-fluoro-2-(1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD24080992
  • Cas Number : 1536670-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726277
Availability details for
Quantity
  • Availability
  • Add to Cart