ENAH11DD2796 Enamine

1-(1-methyl-1H-pyrazol-3-yl)-1H-imidazole-4-carboxylic acid hydrochloride

  • MDL number MFCD31666493
  • Cas Number : 2228674-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726340
Availability details for
Quantity
  • Availability
  • Add to Cart