ENAH11DD27AE Enamine

3-acetyl-5,5-dimethyloxolan-2-one

  • MDL number MFCD22069574
  • Cas Number : 7391-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726424
Availability details for
Quantity
  • Availability
  • Add to Cart