ENAH11DD27B2 Enamine

1,3-thiazole-4-sulfonamide

  • MDL number MFCD18821124
  • Cas Number : 89501-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725011
Availability details for
Quantity
  • Availability
  • Add to Cart