ENAH11DD27B6 Enamine

tert-butyl 4-(1-aminoethyl)-4-hydroxypiperidine-1-carboxylate hydrochloride

  • MDL number MFCD31666717
  • Cas Number : 2228087-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726437
Availability details for
Quantity
  • Availability
  • Add to Cart