ENAH11DD27C3 Enamine

(2S)-1-cyclopropylpropan-2-ol

  • MDL number MFCD23913441
  • Cas Number : 1432621-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725058
Availability details for
Quantity
  • Availability
  • Add to Cart