ENAH11DD27C5 Enamine

methyl({[1-(propan-2-yl)-1H-imidazol-2-yl]methyl})amine

  • MDL number MFCD09908702
  • Cas Number : 1314911-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725065
Availability details for
Quantity
  • Availability
  • Add to Cart