ENAH11DD27CE Enamine

4-amino-1-methyl-2lambda6-thia-1-azaspiro[4.5]dec-3-ene-2,2-dione

  • MDL number MFCD31665418
  • Cas Number : 2088353-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725089
Availability details for
Quantity
  • Availability
  • Add to Cart