ENAH11DD27D4 Enamine

9-azaspiro[bicyclo[3.3.1]nonane-3,4'-imidazolidine]-2',5'-dione hydrochloride

  • MDL number MFCD31666728

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725095
Availability details for
Quantity
  • Availability
  • Add to Cart