ENAH11DD2800 Enamine

2-[(tert-butoxy)carbonyl]-8-oxa-2-azaspiro[4.5]decane-3-carboxylic acid

  • MDL number MFCD31666512
  • Cas Number : 2228087-03-2
  • Molecular Weight : 285.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1725276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725276
Availability details for
Quantity
  • Availability
  • Add to Cart