ENAH11DD2810 Enamine

methyl 4-methylnaphthalene-1-carboxylate

  • MDL number MFCD06203861
  • Cas Number : 35615-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725518
Availability details for
Quantity
  • Availability
  • Add to Cart