ENAH11DD2814 Enamine

methyl 2-amino-1,2,3,4-tetrahydronaphthalene-2-carboxylate hydrochloride

  • MDL number MFCD15142888
  • Cas Number : 1197233-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725523
Availability details for
Quantity
  • Availability
  • Add to Cart