ENAH11DD281B Enamine

2-hydroxy-3-(1-methyl-1H-imidazol-4-yl)propanoic acid hydrochloride

  • MDL number MFCD31665425
  • Cas Number : 2228364-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725538
Availability details for
Quantity
  • Availability
  • Add to Cart