ENAH11DD2839 Enamine

methyl 3-cyano-2-sulfanylidene-1H,2H,5H,6H,7H-cyclopenta[b]pyridine-4-carboxylate

  • MDL number MFCD30005269
  • Cas Number : 2098034-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724775
Availability details for
Quantity
  • Availability
  • Add to Cart