ENAH11DD283A Enamine

2-({11-ethyl-12-oxo-7-thia-9,11-diazatricyclo[6.4.0.0,2,6]dodeca-1(8),2(6),9-trien-10-yl}sulfanyl)acetic acid

  • MDL number MFCD01470980
  • Cas Number : 314041-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725627
Availability details for
Quantity
  • Availability
  • Add to Cart