ENAH11DD283C Enamine

2-(5-methylthiophen-2-yl)morpholine

  • MDL number MFCD11646239
  • Cas Number : 1097820-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725629
Availability details for
Quantity
  • Availability
  • Add to Cart