ENAH11DD750F Enamine

2-(diethoxymethyl)morpholine

  • MDL number MFCD31736048
  • Cas Number : 2228450-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1737467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1737467
Availability details for
Quantity
  • Availability
  • Add to Cart