ENAH11DDEF4C Enamine

3,4,4-trimethylhexanoic acid

  • MDL number MFCD31718415
  • Cas Number : 874007-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1755207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1755207
Availability details for
Quantity
  • Availability
  • Add to Cart