ENAH11DE1043 Enamine

(2R)-1-(2-methyl-2,3-dihydro-1-benzofuran-5-yl)propan-2-amine

  • MDL number MFCD32068542
  • Cas Number : 2227988-71-6
  • Molecular Weight : 191.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1760263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1760263
Availability details for
Quantity
  • Availability
  • Add to Cart