ENAH11DE6BE7 Enamine

6-methoxy-1H-indol-2-ol

  • MDL number MFCD22576569
  • Cas Number : 1935388-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1773994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1773994
Availability details for
Quantity
  • Availability
  • Add to Cart