ENAH11DEDA40 Enamine

5-(oxan-4-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD31718424
  • Cas Number : 1785041-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1790316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1790316
Availability details for
Quantity
  • Availability
  • Add to Cart