ENAH11DF135E Enamine

4,5-difluoro-2-methoxybenzene-1-thiol

  • MDL number MFCD31697407
  • Cas Number : 2168380-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1798192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1798192
Availability details for
Quantity
  • Availability
  • Add to Cart