ENAH11DFB571 Enamine

[1-(oxan-2-yl)cyclopropyl]methanol

  • MDL number MFCD31666461
  • Cas Number : 2153241-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1822578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1822578
Availability details for
Quantity
  • Availability
  • Add to Cart