ENAH11E00B85 Enamine

N-[3-(2-hydroxyethyl)phenyl]acetamide

  • MDL number MFCD30491383
  • Cas Number : 943129-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1840238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1840238
Availability details for
Quantity
  • Availability
  • Add to Cart