ENAH11E0211B Enamine

(2R)-1-cyclopropylpropan-2-ol

  • MDL number MFCD23913442
  • Cas Number : 1432620-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1845765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1845765
Availability details for
Quantity
  • Availability
  • Add to Cart