ENAH11E0239C Enamine

(1S)-1-(2,4,6-trifluorophenyl)ethan-1-ol

  • MDL number MFCD24302727
  • Cas Number : 1568193-05-4
  • Molecular Weight : 176.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1846418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1846418
Availability details for
Quantity
  • Availability
  • Add to Cart