ENAH11E02564 Enamine

3,4,5-trifluorobenzene-1-thiol

  • MDL number MFCD11111065
  • Cas Number : 443683-18-9
  • Molecular Weight : 164.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1846864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1846864
Availability details for
Quantity
  • Availability
  • Add to Cart