ENAH11E034D7 Enamine

2-(2-fluoropyridin-3-yl)ethan-1-ol

  • MDL number MFCD09926577
  • Cas Number : 149489-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1850807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1850807
Availability details for
Quantity
  • Availability
  • Add to Cart