ENAH11E04225 Enamine

3-(4-methoxy-3-methylphenyl)propanoic acid

  • MDL number MFCD00029822
  • Cas Number : 22442-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1854220

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1854220
Availability details for
Quantity
  • Availability
  • Add to Cart