ENAH11E07F00 Enamine

1-methyl-1H-indol-4-ol

  • MDL number MFCD09834885
  • Cas Number : 7556-37-8
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1866261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1866261
Availability details for
Quantity
  • Availability
  • Add to Cart